Tubing cutter for polymer tubing, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
A-327
UPCHA-327EA 131 SGD
UPCHA-327
Tubing cutter for polymer tubing, Upchurch Scientific®
Cutters
Designed to cut polymer tubing ¹/₁₆" and ¹/₈" O.D.
Complete with guide holes to ensure precise cutting. The cutter is durable, reliable and easy to operate.
Delivery information: Complete unit supplied with five replacement blades.