Tubing cutter for polymer tubing, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
A-350
UPCHA-350EA 719 SGD
UPCHA-350
Tubing cutter for polymer tubing, Upchurch Scientific®
Cutters
Designed to cut capillary-sized polymer tubing.
The cutter makes clean, perpendicular cuts without collapsing thin walls. The included tubing sleeves are for cutting 360 µm O.D. polymer capillary tubing. Alternative sleeves are available for cutting 510 µm and ¹/₃₂" ext. Ø tubing. All tubing sleeves are 2" long and are made of FEP.
Delivery information: Supplied with a set of 10 tubing sleeves along with five replacement blades.