Inline check valves, quick-stop luer, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
P-696
UPCHP-696EA 465 SGD
UPCHP-696
Inline check valves, quick-stop luer, Upchurch Scientific®
Valves Non Return Valves
The Quick-Stop Luer check valve assembly is specifically designed to provide inline Luer connect/disconnect convenience while avoiding spills. It is made from biocompatible PEEK with gold-plated stainless steel spring.
  • Remains open when engaged
  • Designed to provide inline Luer connect/disconnect convenience without the mess and hazard of spills