Medium pressure injection valves, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
V-451
UPCHV-451EA 1570 SGD
UPCHV-451
Medium pressure injection valves, Upchurch Scientific®
Valves Injection Valves
The 6-port medium pressure injection valves feature easy, external sample loop customisation. Simply clean-cut a piece of tubing that represents the volume of sample you wish to inject, taking into account the port-to-port volume of the valve.
  • Biocompatible and inert
  • Chemically resistant Vespel® and PTFE resin-wetted surfaces
  • Pressure rated to 1000 psi (69 bar)
Ordering information: Each valve includes six ¹/₄ - 28 flangeless fittings for ¹/₁₆" or ¹/₈" ext. Ø tubing.