Burner head, butterfly design
Supplier: USBECK Laborgeräte
1603
USBE1603EA 15.9 SGD
USBE1603
Burner head, butterfly design
Heaters Gas Burners
Nickel plated brass.
For burner head Ø 17 mm
Width of opening: 50 mm
Order Now
SPECIFICATIONS
Environmentally Preferable