Gas cylinder holder
Supplier: USBECK Laborgeräte
2070
USBE2070EA 146 SGD
USBE2070
Gas cylinder holder
Wall Hooks
Powder coated aluminium gas cylinder holder with safety belt and snap buckles. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 70 mm thickness.
  • Suitable for all cylinders size 10 litres and above
  • Secures tightly, can be loosened quickly