Clamps, universal, slewable
Supplier: USBECK Laborgeräte
2508
USBE2508EA 62.7 SGD
USBE2508
Clamps, universal, slewable
Clamps General Purpose Clamps
Universal clamp made of powder-coated zinc die-cast.With cork lined jaws.
  • Capacity 0 to 80 mm
  • Slewing range 180°