Clamps
Supplier: USBECK Laborgeräte
2574 2564
USBE2574EA 49.5 SGD
USBE2574 USBE2564
Clamps
Clamps General Purpose Clamps
Zinc die-casting, nickel plated.
  • With vinyl coated jaws
  • With rod Ø 10 mm
Order Now
SPECIFICATIONS
Environmentally Preferable