Clamp with rubber belt
Supplier: USBECK Laborgeräte
2825
USBE2825EA 113 SGD
USBE2825
Clamp with rubber belt
Clamps General Purpose Clamps
Rubber belt.
  • Cam type closure
  • Cork lined jaws
  • Capacity Ø 60 - 170 mm
  • Rod Ø 10 mm
Order Now
SPECIFICATIONS
Environmentally Preferable