Pharmacist's spatula, with plastic handle
Supplier: USBECK Laborgeräte
3510 3511 3512 3513 3514
USBE3510EA 13.8 SGD
USBE3510 USBE3511 USBE3512 USBE3513 USBE3514
Pharmacist's spatula, with plastic handle
Spatulas
Highly flexible spatulas with stainless steel blade and plastic handle. Autoclavable.