Instrument trays
Supplier: USBECK Laborgeräte
4760 4762 4764
USBE4760EA 55.5 SGD
USBE4760 USBE4762 USBE4764
Instrument trays
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
Instrument trays with rounded corner.
Order Now
SPECIFICATIONS
Environmentally Preferable