Stands, with or without glass ceramic plate
Supplier: USBECK Laborgeräte
5112 5111 5113 5151 5152 5153 5161 5162 5163
USBE5112EA 43.3 SGD
USBE5112 USBE5111 USBE5113 USBE5151 USBE5152 USBE5153 USBE5161 USBE5162 USBE5163
Stands, with or without glass ceramic plate
Retort Stands
These stainless steel stands with four feet are available with or without glass ceramic plate. They have the optimum height for use with Usbeck laboratory burners.
  • Excellent stability
  • Space saving storage thanks to dismountable feet
  • Glass ceramic plate (thickness 3,8 mm) with chamfered edges can be used above open burner flames
  • Plate holder and plate can also function as a heat resistant table mat