Diamond pen
Supplier: USBECK Laborgeräte
5241
USBE5241EA 87.7 SGD
USBE5241
Diamond pen
Pens Ink Pens
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.