Supplier: ULTRATAPE
1310TB100-P5D
UTAP1310TB100-P5DEA 559 SGD
UTAP1310TB100-P5D
Tapes
UltraTape offers a number of specialty cleanroom tapes that can be customised to meet a wide variety of applications. Special packaging, cores, backing, and adhesive materials are available to meet specific requirements.
  • High temperature, low adhesive wafer processing tapes
  • Customised packaging to meet critical environment requirements
  • Poly TechCut tapes and labels for circuit board mask, wave solder, and other high temperature applications
Tensile strength: 2,45 N/mm
Elongation: 200%