High temperature cleanroom tapes
Supplier: ULTRATAPE
2156BZ050P3D
UTAP2156BZ050P3DEA 76 SGD
UTAP2156BZ050P3D
High temperature cleanroom tapes
Tapes
These high temperature cleanroom tapes are made of polyimide with silicone adhesive.
  • Removes without adhesive residue
  • Resistant to tearing and punctures
  • For use in cleanroom
Length: 32,9 m
Colour: Bronze