Tape dispenser, single roll
Supplier: ULTRATAPE
UT922
UTAPUT922EA 183 SGD
UTAPUT922
Tape dispenser, single roll
Label and Tape Dispensers
UltraTape single roll dispenser is designed to hold one roll up to 50,8 mm width and 201,2 metres in length. The dispenser is supplied with a 76 mm plastic core holder.