Tape dispenser, multiple roll
Supplier: ULTRATAPE
UT924
UTAPUT924EA 606 SGD
UTAPUT924
Tape dispenser, multiple roll
Label and Tape Dispensers
This multiple roll dispenser is designed to hold four rolls of up to 25,4 mm width and 201,2 metres in length.
Delivery information: Supplied with four 76 mm plastic core holders.