Disposable earplugs, hi-com
Supplier: UVEX
2112.100
UVEX2112.100EA 146 SGD
UVEX2112.100
Disposable earplugs, hi-com
Earplugs Disposable Earplugs
The innovative shape of the hi-com reduces the level of sound that can build up in the inner ear. There is no risk of overprotection in areas with medium noise exposure.
  • Earplug design make it possible to understand speech therefore improving communication
  • Available in two colours
  • With low sound absorption
SNR: 24 dB
Certifications: EN 352-2