Safety spectacles, uvex meteor
Supplier: UVEX
9134.005
UVEX9134.005EA 40.6 SGD
UVEX9134.005
Safety spectacles, uvex meteor
Safety Glasses
The dual lens glasses with close fitting frame provide reliable protection and protection against particles from below.
  • Anatomically shaped nose bridge
  • Side shields and upper eye rim protection integrated in frame design
  • Side arms individually adjustable in four different lengths
  • Adjustable comfortable soft-form ear pieces
Certifications: EN 166
Delivery information: Supplied with case and microfibre cloth.