Safety spectacles, uvexuvex i-vo
Supplier: UVEX
9160.265 9160.120
UVEX9160.265EA 46.2 SGD
UVEX9160.265 UVEX9160.120
Safety spectacles, uvexuvex i-vo
Safety Glasses
These modern safety glasses offer exceptional comfort through duo-component technology - the combination of two materials (hard and soft). They combine optimum protection and fit with maximum comfort for the wearer.
  • High level of protection from mechanical and optical hazards, due to high impact resistant polycarbonate lenses
  • Indirect ventilation for maximum freedom from misting and increased safety
  • Angle and length of side pieces are individually adjustable for a comfortable and secure fit
Weight: 51 g
EN 166-168, EN 170; (EN 172: Anti-UV 5-2,5); 1More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable