Cover spectacles, uvex 9161
Supplier: UVEX
9161.005
UVEX9161.005EA 29.4 SGD
UVEX9161.005
Cover spectacles, uvex 9161
Safety Glasses
Cover glasses (OTG) with an unrestricted lateral field of vision can be worn over most prescription spectacles.
  • Indirect ventilation
  • Four position arm length adjustment
  • Ear piece pads guarantee an excellent, pressure-free fit
  • Optidur NHC coating for high scratch and chemical resistance
Weight: 56 g
EN 166-168, EN 170; 1F
Certifications: EN 166, EN 170; 1F