Cover spectacles, uvex super OTG
Supplier: UVEX
9169.065 9169.080 9169.260
UVEX9169.065EA 43.6 SGD
UVEX9169.065 UVEX9169.080 UVEX9169.260
Cover spectacles, uvex super OTG
Safety Glasses
Super OTG cover spectacles have thin, hingeless side arms with Grilamid® (a combination of hard and soft plastics). They combine the benefits of temple glasses with the comfort of a headband.
  • Grilamid® has high impact resistance, even at low temperatures, and is also resistant to chemicals
  • The arms are quick and easy to adjust; arm ends in particular are smooth and flexible
  • Lens inclination of 11° guarantees good visibility when looking down and helps prevent the wearer from tripping
  • Curved lens surface provides excellent coverage of the eye area without
More Product Information