Cleanroom cover spectacles, autoclavable, super f OTG CR
Supplier: UVEX
9169.500
UVEX9169.500EA 59.1 SGD
UVEX9169.500
Cleanroom cover spectacles, autoclavable, super f OTG CR
Safety Glasses
This autoclavable eye protection offers best wearers comfort, highest functionality and an innovative design. Suitable for use over prescription spectacles.
  • Very light autoclavable cover spectacle with inner anti-fog effect
  • Autoclavable at least 10 times at 121 °C for 20 min.
  • Non allergenic material which is free of harmful substances
  • Lens inclination of 11° ensures good view downwards and helps protect the wearer from accidents caused by stumbling
  • Ultra-modern lens technology and extremely flexible and soft ear pieces
Ideal for use in cleanrooms andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable