uvex i-3 Safety Spectacles
Supplier: UVEX
9190.275 9190.280 6108.210 9190.281 9190.880 9190.885
UVEX9190.275EA 45.9 SGD
UVEX9190.275 UVEX9190.280 UVEX6108.210 UVEX9190.281 UVEX9190.880 UVEX9190.885
uvex i-3 Safety Spectacles
Safety Glasses
These innovative safety spectacles are designed for a perfect fit.
  • Flexible side arms with variable inclination
  • Integrated Softflex zone ensures comfort for all head sizes
  • Soft nosepiece, earpieces and Softflex zone at temples ensure a comfortable and secure fit
  • Model i-3 add: Prescription spectacles with integrated dioptre correction for near vision
Certifications: EN 166, EN 170.