Goggles, uvex u-sonic
Supplier: UVEX
9308.247
UVEX9308.247EA 48.5 SGD
UVEX9308.247
Goggles, uvex u-sonic
Goggles
The ergonomic design with flexible soft component means the uvex u-sonic fits perfectly to every face its low weight and low pressure fit make them very comfortable to wear.
  • Combines hard and soft components which adapt to the wearer’s face creating soft, effective seal around the eyes
  • Fully adjustable, high quality head strap offers pressure-free fit
  • Prescription insert possible
  • Compact size and soft materials aid compatibility with respirators and other PPE items
  • Innovative ventilation system ensures pleasant comfortable interior environment
  • Large
More Product Information