Safety helmets, uvex airwing B-S
Supplier: UVEX
9762.521
UVEX9762.521EA 32 SGD
UVEX9762.521
Safety helmets, uvex airwing B-S
Head Protection Safety Helmets
Polyethylene helmet shell with three variable air vents and conventional suspension harness for a comfortable fit.
  • Shorter brim for a wider field of upward vision
  • Provides protection even in extremely low temperatures
Size: 510 to 610 mm
Certifications: EN 397