Leak detection additives, Reveal™
Supplier: UVP ULTRA VIOLET PRODUCTS
96-0150-03
UVPA96-0150-03EA 415 SGD
UVPA96-0150-03
Leak detection additives, Reveal™
Spill Control Materials
This leak detection system brings leaks to light with an ultraviolet lamp and specially formulated fluorescent tracing additives.
  • Locating all leaks at once, even multiple small leaks in the same system
  • Tracer additives work easily and quickly in any system using water, oil, ATF, fuel or refrigerant