Imaging station, ColonyDoc-It™
Supplier: UVP ULTRA VIOLET PRODUCTS
97-0539-02
UVPA97-0539-02EA 9390 SGD
UVPA97-0539-02
Imaging station, ColonyDoc-It™
Colony Counters
The innovative and compact design of the ColonyDoc-It™ enables automated, fast and accurate colony counting. The high resolution DSLR colour camera is specifically designed to capture white light and fluorescent marked colonies and a wide array of samples. The sophisticated yet intuitive system offers researchers easy detection and analysis of media. When colony samples require different light sources, the ColonyDoc-It™ enables the easy selection of bright LED lighting. The epi blue light and optional GFP filter enable users to visualise GFP fluorescence detection. The high resolutionMore Product Information