VACUU.LAN® local vacuum network
Supplier: VACUUBRAND
20677101 20677102 20677104 20677105 20677106 20677107 20677155 20677156 20677175 20677176 20677180 20677182 20677190 20677191 20636668 20677208 20677292 20677293
VACU20677101EA 1540 SGD
VACU20677101 VACU20677102 VACU20677104 VACU20677105 VACU20677106 VACU20677107 VACU20677155 VACU20677156 VACU20677175 VACU20677176 VACU20677180 VACU20677182 VACU20677190 VACU20677191 VACU20636668 VACU20677208 VACU20677292 VACU20677293
VACUU.LAN® local vacuum network
Valves Vacuum Valves
The VACUU.LAN local vacuum network connects several workstations to one vacuum pump. The various modules make it possible to connect almost all popular vacuum applications such as rotary evaporators, vacuum concentrators, gel dryers, drying ovens and filter systems. It can be integrated into new laboratory furniture at the time of assembly or subsequently installed in existing laboratories.
  • Individual connection configuration, vacuum connections to suit the requirements of the workstation
  • Integrated non-return valves to prevent cross-contamination, even
More Product Information