Vacuum gauge, DVR 2pro
Supplier: VACUUBRAND
20682906
VACU20682906EA 2870 SGD
VACU20682906
Vacuum gauge, DVR 2pro
Pumps Vacuum Gauges
The DVR 2pro is a fully electronic, versatile vacuum gauge for the measuring range from atmospheric pressure to 1 mbar.
  • Improved functionality within the navigation menu helps to save energy and to increase the life cycle of the battery
  • Display with digital measuring values alows to detect tendencies and makes it easier to readout precisely
  • Pressure unit user-selectable in mbar, hPa or Torr prevents unnecessary conversions
  • Different vacuum connections and the additional support rod allow the use in almost every laboratory application