Vacuum controller
Supplier: VACUUBRAND
20683160
VACU20683160EA 0 SGD
VACU20683160
Vacuum controller
Pumps Vacuum Controllers
Model CVC 3000: Fully electronic, highly versatile vacuum gauge and controller. The controller is equipped with a built-in capacitive absolute-pressure transducer made from alumina/ceramic as well as a built-in venting valve and switched outlets for a suction line valve and a coolant water valve. It eases work considerably and enables environmentally friendly operation whilst protecting samples, solvent recovery, reduced foam formation and superheating. The optimum vacuum setting can be found quickly and simply, even with unknown mixtures and pressures.
  • Process vacuum system, cooling
More Product Information