Vacuum gauge, DCP 3000
Supplier: VACUUBRAND
20683170
VACU20683170EA 5160 SGD
VACU20683170
Vacuum gauge, DCP 3000
Pumps Vacuum Gauges
Versatile, accurate and robust vacuum gauge. Outstanding measurement accuracy thanks to its capacitive, anti-corrosive gauge head with excellent long-term stability and effective temperature compensation. With its robust plastic housing and splash-proof gauge head, the VSK 3000 is ideal for tough applications.
  • Direct measurement on the system with VACUU BUS technology; up to four external gauge heads can be attached with max. distance of 30 m
  • Large, illuminated screen with analogue display and digital measurement values
  • Measurement values of all sensors
More Product Information