Chemistry vacuum pumps and systems MD 1C
Supplier: VACUUBRAND
20696602
VACU20696602EA 10990 SGD
VACU20696602
Chemistry vacuum pumps and systems MD 1C
Pumps Vacuum Diaphragm Pumps
These chemistry vacuum systems can be used in a wide range of applications for evacuating or moving gases and vapours between containers in chemistry and physics laboratories, e.g. as an alternative to water jet pumps and rotary vane pumps. Compact construction, very quiet running, long service life; diaphragm and valves are very easy to replace. Oil-free vacuum.
  • Includes chemistry diaphragm pump with high chemical resistance (parts in contact with media are made from PTFE compounds and similar)
  • High pump volume capacity even at low pressures
  • Standard
More Product Information