Kjeldahl weighing boat
Supplier: VELP SCIENTIFIC
CM0486000
VELPCM0486000EA 619 SGD
VELPCM0486000
Kjeldahl weighing boat
Boats Weighing Boats
Weighing boat is suitable for the syrupy and viscose substances, pulps and powders.
  • Nitrogen-free boat, ensures low error risk
  • Suitable for quantitative transfer of substance into mineralisation test tubes