BOD Incubators, FOC Series
Supplier: VELP SCIENTIFIC
F10300305
VELPF10300305EA 5730 SGD
VELPF10300305
BOD Incubators, FOC Series
Incubators BOD Incubators
These BOD incubators have been designed to keep any product at a constant temperature, where accurate and precise temperature control is required. For applications where monitoring the sample is extremely important, the I models are ideal. The controlled heating and cooling thermoregulation system enables the user to select an operating temperature between 3 and 50 °C. The E models have wireless connection for TEMPSoft™. The I models have an internal transparent door and wireless connection for TEMPSoft™ and an auto tuning system which provides constant control of the internal temperature,More Product Information