Overhead stirrers, electronic, OHS digital and advance
Supplier: VELP SCIENTIFIC
F20100493 F20100494 F20100480 F20100481 F20100482 F20100490 F20100491 F20100492
VELPF20100493EA 4200 SGD
VELPF20100493 VELPF20100494 VELPF20100480 VELPF20100481 VELPF20100482 VELPF20100490 VELPF20100491 VELPF20100492
Overhead stirrers, electronic, OHS digital and advance
Stirrers Overhead Stirrers
The OHS Series overhead stirrers are the ideal solution for the most demanding applications: The powerful brushless motor and microprocessor ensure excellent performance and make the VELP OHS Series overhead stirrers unlike any conventional stirrer.
  • Premium stirring power to complete any task
  • Brushless motor ensures maximum power up to 200 Ncm
  • OHS series maintain a constant torque at any speed (up to 2000 rpm)
  • Torque indicator to monitor viscosity changes
  • Integral timer
  • SpeedServo™ counter-reaction technology
  • Models with single- and double-speed gears