Multi-position magnetic stirrers, AMI series
Supplier: VELP SCIENTIFIC
F204A0167 F204A0168
VELPF204A0167EA 1390 SGD
VELPF204A0167 VELPF204A0168
Multi-position magnetic stirrers, AMI series
Stirrers Magnetic Stirrers, Multi-position
Illuminated magnetic stirrers with single-position (AMI) or four separately controlled positions (AMI 4). Specially designed for titrations, and particularly for those in which optimum lighting conditions are needed in order to see the final point of colour change well. Particularly recommended for titrations that have weak colour changes during conversion.
  • High-power driving magnet type 'PCM' operated by a mono-phase motor for continuous operation
  • Chemically resistant housing
  • Excellent speed control even at low revolutions