Magnetic stirrer, ESP
Supplier: VELP SCIENTIFIC
F206A0179
VELPF206A0179EA 1390 SGD
VELPF206A0179
Magnetic stirrer, ESP
Stirrers Magnetic Stirrers
Ultraflat magnetic stirrer without moving mechanical components.
  • Microprocessor controlled adjustable stirring system
  • Gentle start-up allows optimum progression of stirring speed
  • Plate and housing made of chemically resistant materials