Sterilisation flat reels, Stericlin®
Supplier: VP Medical Packaging
3FKFS230114 3FKFS230110 3FKFS230112 3FKFS230116
VEPA3FKFS230114EA 406 SGD
VEPA3FKFS230114 VEPA3FKFS230110 VEPA3FKFS230112 VEPA3FKFS230116
Sterilisation flat reels, Stericlin®
Bags Sterilisation Pouches
These see-through sterilisation reels made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certifications: DIN EN ISO 11607 and DIN 868-5.
Order Now
SPECIFICATIONS
Environmentally Preferable