Sterilisation gusseted reels, Stericlin®
Supplier: VP Medical Packaging
3FKSS230214
VEPA3FKSS230214EA 339 SGD
VEPA3FKSS230214
Sterilisation gusseted reels, Stericlin®
Bags Sterilisation Pouches
These see-through sterilisation reels made of sterilisation proof 70 g/m2 heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certifications: DIN EN ISO 11607 and DIN 868-5.