Silicone stoppers, Versilic®
Supplier: Saint Gobain Life Sciences
261034
VERN261034EA 82 SGD
VERN261034
Silicone stoppers, Versilic®
Stoppers Silicone Stoppers
These slightly conical translucent stoppers are made from silicone.
  • Non-toxic
  • Biologically neutral, good chemical resistance
  • Odourless and tasteless
  • Resistant to ageing, ozone and repeated sterilisation
Temperature resistant from −60 to +200 °C (briefly up to +220 °C).