Silicone stoppers with folding skirt, Versilic®
Supplier: Saint Gobain Life Sciences
467010-50 467015-20 467020-20 467025-10 467030-05 467018-20 467013-50
VERN467010-50EA 62 SGD
VERN467010-50 VERN467015-20 VERN467020-20 VERN467025-10 VERN467030-05 VERN467018-20 VERN467013-50
Silicone stoppers with folding skirt, Versilic®
Stoppers Silicone Stoppers
These stoppers are made of translucent silicone. The stoppers guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers are imperfect.
  • Autoclavable
  • Outstanding sealing and protection from the outside
  • Biologically neutral, good chemical resistance
  • Resistant to ageing, ozone and repeated sterilisation
  • Temperature resistant from –60 to +220 °C (up to +250 °C for short periods)
The stoppers can be punctured and resealed after sampling, even after several uses.
Resistant to temperatures from -60 to +220 °CMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable