PVF fluoropolymer gas and liquid sampling bags
Supplier: Saint Gobain Life Sciences
D1075011
VERND1075011EA 196 SGD
VERND1075011
PVF fluoropolymer gas and liquid sampling bags
Bags Gas Bags
Made from 2 mil thick PVF fluoropolymer film. When the chemical inertness of FEP is not required, Chemware® heat-sealed, PVF gas sampling bags are the ideal choice. These reusable bags are the perfect economical alternative for analysis of non-corrosive gases. Can be used when the user is dealing with carbon monoxide, sulphur dioxide, hydrogen sulfide, radon and mercaptans. Ideal for extracting air samples from smoke stack exhaust systems in fuel gas sampling applications.
  • Tough, easy to use, reliable
  • Highly flexible – easy to handle and store
  • Leak proof,
More Product Information