Tubing, Tygon® 3350
Supplier: Saint Gobain Life Sciences
T3304-13 T330G-203 T3302-13 T3302-23 T3304-23 T3308-23 T3310-23 T3306-33 T3308-43 T3316-43 T3303-13 T3306-13 T3308-13 T3312-33 T3314-23 T3324-43 T3320-43 T3305-13 T330N-203 T330P-203 T3310-33 T3303-23 T330B-103 T330H-153 T330L-153 T3312-43 T3306-23 T3312-23 T3316-33 T3314-33
VERNT3304-13EA 245 SGD
VERNT3304-13 VERNT330G-203 VERNT3302-13 VERNT3302-23 VERNT3304-23 VERNT3308-23 VERNT3310-23 VERNT3306-33 VERNT3308-43 VERNT3316-43 VERNT3303-13 VERNT3306-13 VERNT3308-13 VERNT3312-33 VERNT3314-23 VERNT3324-43 VERNT3320-43 VERNT3305-13 VERNT330N-203 VERNT330P-203 VERNT3310-33 VERNT3303-23 VERNT330B-103 VERNT330H-153 VERNT330L-153 VERNT3312-43 VERNT3306-23 VERNT3312-23 VERNT3316-33 VERNT3314-33
Tubing, Tygon® 3350
Tubing Transfer Tubing
Silicone, platinum cured, translucent.
  • Biocompatible, ideal for sensitive applications, can be used for foodstuffs
  • Resistant to ozone and UV, autoclavable
Tygon® tubing made from platinum-cured silicone with ultra-smooth surface. High elasticity with good restoring force for constant liquid transfer, even with intermittent pumping action.
Temperature resistance: −80 to +204 °C
Hardness (Shore durometer A): 50
Density: 1,14
Certifications: FDA approved, compliant with USP class VI.
Delivery information: Tubing is double bagged inside aMore Product Information