Safety wash bottles, narrow mouth, printed, VITsafe™
Supplier: VITLAB
1332819 1332839 1332849 1332859 1332869 1332879 1332889 1332899 1332909 1332949 1332959 1332969 1332979 1331819 1331839 1331849 1331859 1331869 1431829 1432829 1432989 1333839 1333849 1333859 1333869 1433829
VITL1332819EA 25 SGD
VITL1332819 VITL1332839 VITL1332849 VITL1332859 VITL1332869 VITL1332879 VITL1332889 VITL1332899 VITL1332909 VITL1332949 VITL1332959 VITL1332969 VITL1332979 VITL1331819 VITL1331839 VITL1331849 VITL1331859 VITL1331869 VITL1431829 VITL1432829 VITL1432989 VITL1333839 VITL1333849 VITL1333859 VITL1333869 VITL1433829
Safety wash bottles, narrow mouth, printed, VITsafe™
Bottles Wash Bottles
LDPE, with VENT CAP threaded connection.
  • Colour coded contents with hazard symbols and descriptions of hazards as required by the Hazardous Substance Ordinance
  • Good chemical resistance
  • Good thermal resistance
VENT CAP works on the principle of gas/vapour permeability. The screw cap prevents any build-up of vapour pressure above the contents. This prevents the risk of solvent dripping or leaking.
Caution: Note: Washbottle 'Hexan' is not suitable for n-Hexane.
Order Now
SPECIFICATIONS
Environmentally Preferable