Gas washing bottles
Supplier: VITLAB
159697 159497 159597
VITL159697EA 2220 SGD
VITL159697 VITL159497 VITL159597
Gas washing bottles
Bottles Gas Washing Bottles
PFA, translucent.
  • Especially suitable for use in trace analysis
  • Long term maintenance of low-concentration reference materials in the PFA containers
  • No memory effects
  • Practically no carryover, no cross-contamination due to the extremely hydrophobic, anti-adhesive, smooth surfaces
  • High thermal stability, from -200 °C to +260 °C, autoclavable
  • Chemical inertness against nearly all chemicals
  • Easy to clean
Cap with S 40 buttress threads, and frit made of PTFE. A pore size of approx. 50 μm for optimal optimal pearling of the gas into the liquid. A wideMore Product Information