Pipette controller, VITLAB maneus®
Supplier: VITLAB
1630500
VITL1630500EA 201 SGD
VITL1630500
Pipette controller, VITLAB maneus®
Pipettes Pipette Controllers
The VITLAB maneus® pipette controller enables both left- and right-handers to operate all normal bulb and graduated pipettes easily and fatigue-free. For use with all normal bulb and graduated pipettes from 0,1 to 200 ml.
  • Safe and easy handling allows even inexperienced users to adjust the meniscus precisely
  • Easy and fast replacement of the hydrophobic membrane filter
  • Optimised valve system: Liquids can be drawn up simply, without exerting pressure
  • Suction element provides rapid filling of the pipette (capacity: 50 ml in less than 10 seconds)
  • Discharge
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable