Tray, with lid
Supplier: VITLAB
43610
VITL43610EA 50.1 SGD
VITL43610
Tray, with lid
Trays/Dishes/Pots Laboratory Trays/Dishes
Rectangular tray, made of white PP with lid, ideal for a storage applications.
  • Easy to clean
  • Broad, stable, easy to grip edge
  • Smooth surface, rounded corners and edges