Volumetric flasks, class A
Supplier: VITLAB
67604
VITL67604EA 60.7 SGD
VITL67604
Volumetric flasks, class A
Flasks Volumetric Flasks
PMP (TPX®), transparent, with PP stopper.
  • Individually calibrated ring mark
  • Imprinted lot number
  • Complete with certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable