Volumetric flasks, class B, red markings
Supplier: VITLAB
676895
VITL676895EA 60.4 SGD
VITL676895
Volumetric flasks, class B, red markings
Flasks Volumetric Flasks
PMP, crystal clear, with screw cap.
  • With ring mark individually calibrated to 'In'
  • Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
  • To preserve markings. cleaning at no higher than 60 °C is recommended
Certifications: Class B tolerances according to DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable